missing translation for 'onlineSavingsMsg'
Learn More

3-Benzyloxyphenyl isothiocyanate, 97%

Product Code. 11344116
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11344116 250 mg 250mg
11334116 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11344116 Supplier Thermo Scientific Alfa Aesar Supplier No. L19242.MD
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 206559-36-6 | C14H11NOS | 241.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 206559-36-6
Molecular Formula C14H11NOS
Molecular Weight (g/mol) 241.31
MDL Number MFCD00060374
InChI Key QJLFNDXMZQJMDL-UHFFFAOYSA-N
Synonym 3-benzyloxyphenyl isothiocyanate, 3-benzyloxyphenylisothiocyanate, 1-benzyloxy-3-isothiocyanatobenzene, acmc-20an9f, 3-phenylmethoxy benzenisothiocyanate, 1-benzyloxy-3-isothiocyanatobenzene #, 1-isothiocyanato-3-phenylmethoxy-benzene, benzene, 1-isothiocyanato-3-phenylmethoxy, benzene, 1-isothiocyanato-3-phenylmethoxy-9ci
PubChem CID 561446
IUPAC Name 1-isothiocyanato-3-phenylmethoxybenzene
SMILES S=C=NC1=CC(OCC2=CC=CC=C2)=CC=C1

Specifications

Melting Point 49°C to 51°C
Quantity 250 mg
UN Number UN2923
Sensitivity Moisture sensitive
Formula Weight 241.31
Percent Purity 97%
Chemical Name or Material 3-Benzyloxyphenyl isothiocyanate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.