missing translation for 'onlineSavingsMsg'
Learn More

3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic acid, 99%

Product Code. 15422536
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15422536 250 g 250g
15412536 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15422536 Supplier Thermo Scientific Alfa Aesar Supplier No. H56187.30
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 68399-80-4 | C7H17NO6S | 243.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-80-4
Molecular Formula C7H17NO6S
Molecular Weight (g/mol) 243.274
MDL Number MFCD00038353
InChI Key XCBLFURAFHFFJF-UHFFFAOYSA-N
Synonym dipso, 3-bis 2-hydroxyethyl amino-2-hydroxypropane-1-sulfonic acid, 3-n,n-bis 2-hydroxyethyl amino-2-hydroxypropanesulfonic acid, 3-n,n-bis 2-hydroxyethyl amino-2-hydroxy-1-propanesulfonic acid, 1-propanesulfonic acid, 3-bis 2-hydroxyethyl amino-2-hydroxy, 3-bis 2-hydroxyethyl amino-2-hydroxypropanesulfonic acid, 3-bis 2-hydroxyethyl amino-2-hydroxy-1-propanesulfonic acid, n,n-bis 2-hydroxyethyl-3-amino-2-hydroxypropanesulfonic acid
PubChem CID 100210
IUPAC Name 3-[bis(2-hydroxyethyl)amino]-2-hydroxypropane-1-sulfonic acid
SMILES C(CO)N(CCO)CC(CS(=O)(=O)O)O

Specifications

Quantity 250 g
Beilstein 2105454
Formula Weight 261.29
Percent Purity 99%
Chemical Name or Material 3-[Bis(2-hydroxyethyl)amino]-2-hydroxypropanesulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.