missing translation for 'onlineSavingsMsg'
Learn More

3-(BOC-amino)-1-propanol, 97%

Product Code. 10214684 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 mL
50 mL
Unit Size:
10mL
50mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10214684 50 mL 50mL
10143394 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10214684 Supplier Thermo Scientific Acros Supplier No. 399800500
 Restricted Item
Estimated Shipment: 01-10-2026
Add to basket
This item is not returnable. View return policy

CAS: 58885-58-8 | C8H17NO3 | 175.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58885-58-8
Molecular Formula C8H17NO3
Molecular Weight (g/mol) 175.23
MDL Number MFCD00191883
InChI Key XDJCYKMWJCYQJM-UHFFFAOYSA-N
Synonym 3-boc-amino-1-propanol, tert-butyl n-3-hydroxypropyl carbamate, boc-beta-ala-ol, 3-tert-butoxycarbonylamino-1-propanol, tert-butyl 3-hydroxypropylcarbamate, 3-n-boc-amino propanol, tert-butyl 3-hydroxypropyl carbamate, n-3-hydroxypropyl carbamic acid tert-butyl ester, 3-hydroxy-propyl-carbamic acid tert-butyl ester
PubChem CID 3838134
IUPAC Name tert-butyl N-(3-hydroxypropyl)carbamate
SMILES CC(C)(C)OC(=O)NCCCO

Specifications

Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Linear Formula HO(CH2)3NHCO2C(CH3)3
Density 1.0250g/mL
Quantity 50 mL
Flash Point 109°C
Formula Weight 175.23
Percent Purity 97%
Specific Gravity 1.025
Chemical Name or Material 3-(BOC-amino)-1-propanol
compliance-icons
Product Identifier
  • 3-(BOC-amino)-1-propanol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.