missing translation for 'onlineSavingsMsg'
Learn More

3-(Boc-amino)cyclohexanecarboxylic acid, 98+%

Product Code. 15448927
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448927 1 g 1g
15468927 250 mg 250mg
15458927 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15448927 Supplier Thermo Scientific Alfa Aesar Supplier No. H52299.03
 Restricted Item
Estimated Shipment: 03-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 334932-13-7 | C12H21NO4 | 243.303 g/mol

3-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-(Boc-amino)cyclohexanecarboxylic acid is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water (2.1 g/L at 25°C).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 334932-13-7
Molecular Formula C12H21NO4
Molecular Weight (g/mol) 243.303
MDL Number MFCD03453252
InChI Key JSGHMGKJNZTKGF-UHFFFAOYSA-N
Synonym 3-tert-butoxycarbonyl amino cyclohexanecarboxylic acid, 3-boc-amino cyclohexanecarboxylic acid, 3-tert-butoxy carbonyl amino cyclohexane-1-carboxylic acid, 3-2-methylpropan-2-yl oxycarbonylamino cyclohexane-1-carboxylic acid, cyclohexanecarboxylic acid, 3-1,1-dimethylethoxy carbonyl amino, cis-3-boc-amino cyclohexanecarboxylic acid, trans-3-tert-butoxycarbonylamino cyclohexanecarboxylic acid, 3-tert-butoxycarbonyl amino cyclohexane-1-carboxylic acid, acmc-209fp1
PubChem CID 4420316
IUPAC Name 3-[(2-methylpropan-2-yl)oxycarbonylamino]cyclohexane-1-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1CCCC(C1)C(=O)O

Specifications

Solubility Information Slightly soluble in water (2.1g/L at 25°C).
Quantity 1 g
Formula Weight 243.3
Percent Purity ≥98%
Chemical Name or Material 3-(Boc-amino)cyclohexanecarboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.