missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-butanone, 95%, stabilized

Product Code. 10687654 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
Product Code. Quantity Packaging unitSize
10687654 1 g Glass bottle 1g
10381581 5 g Glass bottle 5g
2 options
This item is not returnable. View return policy

Product Code. 10687654

Brand: Thermo Scientific Acros 402910010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 814-75-5 | C4H7BrO | 151 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-75-5
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151
MDL Number MFCD00013538
InChI Key BNBOUFHCTIFWHN-UHFFFAOYSA-N
Synonym 3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on
PubChem CID 13142
IUPAC Name 3-bromobutan-2-one
SMILES CC(C(=O)C)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color Yellow to Brown
Density 1.4380g/mL
Boiling Point 36.0°C (11.0 mmHg)
Flash Point 51°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula CH3COCHBrCH3
Refractive Index 1.4580 to 1.4600
Quantity 1 g
Specific Gravity 1.438
Stabilizer 1% max. magnesium oxide
Formula Weight 151
Percent Purity 95%
Physical Form Liquid
Chemical Name or Material 3-Bromo-2-butanone, 95%
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Bromo-2-butanone, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.