missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-butanone, 97%, stab. with <1% magnesium oxide

Product Code. 11419492
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
Unit Size:
10g
250g
50g
Product Code. Quantity unitSize
11419492 10 g 10g
11429492 50 g 50g
11439492 250 g 250g
3 options
This item is not returnable. View return policy

Product Code. 11419492

Brand: Thermo Scientific Alfa Aesar A10186.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 814-75-5 | C4H7BrO | 151.003 g/mol

3-Bromo-2-butanone is involved in the preparation of o-isopropyl S-3-oxobutan-2-yl dithiocarbonate by reaction with potassium o-isopropylxanthate, which on treatment with sulfuric acid gives 4,5-dimethyl-1,3-dithiol-2-one .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-2-butanone is involved in the preparation of o-isopropyl S-3-oxobutan-2-yl dithiocarbonate by reaction with potassium o-isopropylxanthate, which on treatment with sulfuric acid gives 4,5-dimethyl-1,3-dithiol-2-one .

Solubility
Miscible with dichloromethane.

Notes
Incompatible with strong oxidizing agents, strong bases and strong reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 814-75-5
Molecular Formula C4H7BrO
Molecular Weight (g/mol) 151.003
MDL Number MFCD00013538
InChI Key BNBOUFHCTIFWHN-UHFFFAOYSA-N
Synonym 3-bromo-2-butanone, 2-butanone, 3-bromo, 2-bromo-3-butanone, 3-bromo-butan-2-one, 1-bromoethyl methyl ketone, 2-bromobutan-2-one, 3-bromobutanone, 3-bromo-2-butanon, 2-bromo-3-oxobutane, 3-bromo-butan-2-on
PubChem CID 13142
IUPAC Name 3-bromobutan-2-one
SMILES CC(C(=O)C)Br

Specifications

Density 1.438
Boiling Point 69°C to 70°C (70 mmHg)
Flash Point >100°C (212°F)
Odor Pungent
Refractive Index 1.459
Quantity 10 g
UN Number UN2922
Beilstein 906773
Solubility Information Miscible with dichloromethane.
Formula Weight 151.01
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-butanone, Stabilized with <1% magnesium oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.