missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-furoyl chloride, 97%, Thermo Scientific™

Product Code. 10739025
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10739025 5 g Amber Glass Bottle 5g
10163464 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10739025 Supplier Thermo Scientific Maybridge Supplier No. CC67202DE
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 58777-57-4
Molecular Formula C5H2BrClO2
Molecular Weight (g/mol) 209.423
MDL Number MFCD11109330
InChI Key WTZDPJVRZQGTET-UHFFFAOYSA-N
Synonym 3-bromo-2-furoyl chloride, 3-bromo-2-furancarbonyl chloride, 3-bromo-furan-2-carbonyl chloride, 2-furancarbonyl chloride, 3-bromo-9ci
PubChem CID 21864576
IUPAC Name 3-bromofuran-2-carbonyl chloride
SMILES C1=COC(=C1Br)C(=O)Cl

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 209.43
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-furoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.