missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methyl-1-propene, TRC

Product Code. 30337464
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30337464 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30337464 Supplier Toronto Research Chemicals Supplier No. TRCB68548025G
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromo-2-methyl-1-propene
CAS 1458-98-6
Molecular Formula C4H7Br
Quantity 25 g
Synonym 3-bromo-2-methyl-propene, 1-Bromo-2-methyl-2-propene, 2-(Bromomethyl)propene, 2-Methyl-2-propenyl Bromide, 2-Methyl-3-bromopropene, 2-Methylallyl bromide, 3-Bromo-2-methyl-1-propene, 3-Bromo-2-methyl-1-propylene, 3-Bromo-2-methylpropene, 3-Bromoisobutylene, Methallyl Bromide, Methylallyl Bromide, NSC 617310, beta-Methallyl Bromide, beta-Methylallyl Bromide
InChI Formula InChI=1S/C4H7Br/c1-4(2)3-5/h1,3H2,2H3
SMILES CC(=C)CBr
IUPAC Name 3-bromo-2-methylprop-1-ene
Molecular Weight (g/mol) 135
Formula Weight 133.9731
Product Title
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.