missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-2-methylpropene, 97%

Product Code. 10149599 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10149599 5 mL Glass bottle 5mL
10832251 25 mL Glass bottle 25mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10149599 Supplier Thermo Scientific Acros Supplier No. 427650050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1458-98-6 | C4H7Br | 135.00 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1458-98-6
Molecular Formula C4H7Br
Molecular Weight (g/mol) 135.00
MDL Number MFCD00134155
InChI Key USEGQJLHQSTGHW-UHFFFAOYSA-N
Synonym 3-bromo-2-methylpropene, 3-bromo-2-methyl-1-propene, methallyl bromide, 2-bromomethyl prop-1-ene, 2-methylallyl bromide, 1-propene, 3-bromo-2-methyl, 3-bromo-2-methyl-prop-1-ene, 3-bromo-2-methylpropene, stabilized with hydroquinone, methallylbromide, methylallyl bromide
PubChem CID 357785
IUPAC Name 3-bromo-2-methylprop-1-ene
SMILES CC(=C)CBr

Specifications

CAS Min % 96.0
CAS Max % 100.0
Density 1.3390g/mL
Boiling Point 94.0°C to 95.0°C
Flash Point 7°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH2=C(CH3)CH2Br
Quantity 5 mL
Specific Gravity 1.339
Formula Weight 135
Percent Purity 97%
Chemical Name or Material 3-Bromo-2-methylpropene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromo-2-methylpropene
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.