missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromo-4-hydroxybenzonitrile, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 3-Bromo-4-hydroxybenzonitrile |
| CAS | 2315-86-8 |
| Molecular Formula | C7H4BrNO |
| Quantity | 10 g |
| Synonym | 2-Bromo-4-cyanophenol |
| InChI Formula | InChI=1S/C7H4BrNO/c8-6-3-5(4-9)1-2-7(6)10/h1-3,10H |
| SMILES | BrC1=C(O)C=CC(C#N)=C1 |
| IUPAC Name | 3-bromo-4-hydroxybenzonitrile |
| Molecular Weight (g/mol) | 198.02 |
| Formula Weight | 198.95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction