missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-isocyanatothiophene, 95%, Thermo Scientific™

Product Code. 10041384
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10041384 1 g Amber Glass Bottle 1g
10674385 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10041384 Supplier Thermo Scientific Maybridge Supplier No. CC54206DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 937795-88-5
Molecular Formula C5H2BrNOS
Molecular Weight (g/mol) 204.041
MDL Number MFCD09879904
InChI Key HVJDEHSHJHNENX-UHFFFAOYSA-N
Synonym 4-bromothiophen-3-isocyanate, 4-bromothien-3-yl isocyanate
PubChem CID 24229539
IUPAC Name 3-bromo-4-isocyanatothiophene
SMILES C1=C(C(=CS1)Br)N=C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.05
Percent Purity 95%
Chemical Name or Material 3-Bromo-4-isocyanatothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.