missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-isocyanatothiophene, 95%, Thermo Scientific™

Product Code. 10674385
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10674385 5 g Amber Glass Bottle 5g
10041384 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10674385 Supplier Thermo Scientific Maybridge Supplier No. CC54206DE
 Restricted Item
Estimated Shipment: 04-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 937795-88-5
Molecular Formula C5H2BrNOS
Molecular Weight (g/mol) 204.041
MDL Number MFCD09879904
InChI Key HVJDEHSHJHNENX-UHFFFAOYSA-N
Synonym 4-bromothiophen-3-isocyanate, 4-bromothien-3-yl isocyanate
PubChem CID 24229539
IUPAC Name 3-bromo-4-isocyanatothiophene
SMILES C1=C(C(=CS1)Br)N=C=O

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 204.05
Percent Purity 95%
Chemical Name or Material 3-Bromo-4-isocyanatothiophene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.