missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-nitroindole, 97%

Product Code. 15420199
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
Product Code. Quantity unitSize
15420199 5 g 5g
15430199 250 mg 250mg
15410199 1 g 1g
3 options
This item is not returnable. View return policy

Product Code. 15420199

Brand: Thermo Scientific Alfa Aesar H63478.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 126807-08-7 | C8H5BrN2O2 | 241.044 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 126807-08-7
Molecular Formula C8H5BrN2O2
Molecular Weight (g/mol) 241.044
InChI Key VYQRPDVMVMLVHA-UHFFFAOYSA-N
Synonym 3-bromo-4-nitroindole, 1h-indole,3-bromo-4-nitro, acmc-20dlf4, 1h-indole, 3-bromo-4-nitro
PubChem CID 14707933
IUPAC Name 3-bromo-4-nitro-1H-indole
SMILES C1=CC2=C(C(=C1)[N+](=O)[O-])C(=CN2)Br

Specifications

Quantity 5 g
Formula Weight 241.04
Percent Purity 97%
Physical Form Solid
Chemical Name or Material 3-Bromo-4-nitroindole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.