missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-4-pyridinecarboxaldehyde, 97%

Product Code. 10587824 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkode Quantity Packaging Förpackningsstorlek
10587824 1 g Glass bottle 1g
10386684 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denne vare kan ikke returneres. Se returpolitik
Produktkode 10587824 missing translation for 'mfr' Thermo Scientific Acros missing translation for 'supplierNo' 398080010
 Restricted Item
Estimated Shipment: 03-12-2026
Tilføj til indkøbskurv
Tilføj til indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolitik

CAS: 70201-43-3 | C6H4BrNO | 186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 70201-43-3
Molecular Formula C6H4BrNO
Molecular Weight (g/mol) 186
InChI Key NOBDKWLIAQKADB-UHFFFAOYSA-N
Synonym 3-bromoisonicotinaldehyde, 3-bromo-4-pyridinecarboxaldehyde, 3-bromopyridine-4-carboxaldehyde, 3-bromo-4-formylpyridine, 3-bromo-4-pyridinecarbaldehyde, 4-pyridinecarboxaldehyde, 3-bromo, 3-bromo-4-pyridincarbaldehyde, bromoisonicotinaldehyde, pubchem5063, acmc-1blzq
PubChem CID 2762997
IUPAC Name 3-bromopyridine-4-carbaldehyde
SMILES C1=CN=CC(=C1C=O)Br

Tekniske data

Melting Point 80°C to 83°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 186
Percent Purity 97%
Chemical Name or Material 3-Bromo-4-pyridinecarboxaldehyde
compliance-icons
Produktidentifikator
  • 3-Bromo-4-pyridinecarboxaldehyde
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.