missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(chloromethyl)pyridine hydrochloride, 95%, Thermo Scientific™

Product Code. 10547664
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10547664 1 g Amber Glass Bottle 1g
10716915 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10547664 Supplier Thermo Scientific Maybridge Supplier No. CC67174DA
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 39741-46-3
Molecular Formula C6H6BrCl2N
Molecular Weight (g/mol) 242.925
MDL Number MFCD09260933
InChI Key CJAUTVHMRSAZGG-UHFFFAOYSA-N
Synonym 3-bromo-5-chloromethyl pyridine hydrochloride, 3-bromo-5-chloromethyl pyridine hcl, pyridine, 3-bromo-5-chloromethyl-, hydrochloride, 3-bromo-5-chloromethyl-pyridine hcl salt, ksc776i7t, 5-bromo-3-chloromethyl pyridine, chloride, 3-bromo-5-chloromethylpyridine hydrochloride, 3-bromo-5-chloromethyl pyridinehydrochloride, 3-bromo-5-chloromethyl-pyridine hydrochloride, 3-bromo-5-chloromethyl pyridine hydrochloride 1:1
PubChem CID 12515069
IUPAC Name 3-bromo-5-(chloromethyl)pyridine;hydrochloride
SMILES C1=C(C=NC=C1Br)CCl.Cl

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 242.93
Percent Purity 95%
Chemical Name or Material 3-Bromo-5-(chloromethyl)pyridine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.