missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)pyridine, 95%

Product Code. 11455540
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11455540 1 g 1g
11465540 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11455540 Supplier Thermo Scientific Alfa Aesar Supplier No. H25823.03
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 436799-33-6 | C6H3BrF3N | 225.996 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 436799-33-6
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 225.996
MDL Number MFCD04972700
InChI Key HEDHNDVPKRVQPN-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 5-bromo-3-trifluoromethyl pyridine, pyridine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluormethyl-pyridine, abbypharma ap-11-5533, 3-trifluoromethyl-5-bromopyridine, pubchem3010, acmc-209jv5, 3-bromo-5-trifluoromethylpyridin
PubChem CID 11127991
IUPAC Name 3-bromo-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC=C1Br)C(F)(F)F

Specifications

Melting Point 44°C to 46°C
Boiling Point 65°C to 66°C (19 mmHg)
Flash Point 85°C (185°F)
Quantity 1 g
Formula Weight 226
Percent Purity 95%
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)pyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.