missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(trifluoromethyl)pyridine, 97%, Thermo Scientific™

Product Code. 10339747 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10339747 1 g Glass Bottle 1g
10533098 5 g Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10339747 Supplier Thermo Scientific Acros Supplier No. 429890010

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 436799-33-6
Molecular Formula C6H3BrF3N
Molecular Weight (g/mol) 226
MDL Number MFCD04972700
InChI Key HEDHNDVPKRVQPN-UHFFFAOYSA-N
Synonym 3-bromo-5-trifluoromethyl pyridine, 3-bromo-5-trifluoromethyl-pyridine, 5-bromo-3-trifluoromethyl pyridine, pyridine, 3-bromo-5-trifluoromethyl, 3-bromo-5-trifluormethyl-pyridine, abbypharma ap-11-5533, 3-trifluoromethyl-5-bromopyridine, pubchem3010, acmc-209jv5, 3-bromo-5-trifluoromethylpyridin
PubChem CID 11127991
IUPAC Name 3-bromo-5-(trifluoromethyl)pyridine
SMILES C1=C(C=NC=C1Br)C(F)(F)F

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 43.5°C to 46.0°C
Color White to Yellow
Boiling Point 65.0°C to 66.0°C (19.0 mmHg)
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 226
Percent Purity 97%
Physical Form Crystalline Powder or Solid
Chemical Name or Material 3-Bromo-5-(trifluoromethyl)pyridine
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromo-5-(trifluoromethyl)pyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.