missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-7-nitroindazole, 98+%

Product Code. 15495699
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15495699 1 g 1g
15415709 250 mg 250mg
15405709 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15495699 Supplier Thermo Scientific Alfa Aesar Supplier No. J63480.03
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Inhibitor of nitric oxide synthase (NOS) | CAS: 74209-34-0 | C7H4BrN3O2 | 242.03 g/mol

Inhibitor of nitric oxide synthase (NOS)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 74209-34-0
Molecular Formula C7H4BrN3O2
Molecular Weight (g/mol) 242.03
MDL Number MFCD00159910
InChI Key NFSTZPMYAZRZPC-UHFFFAOYSA-N
Synonym 3-bromo-7-nitroindazole, 3-bromo-7-nitro-1h-indazole, 1h-indazole,3-bromo-7-nitro, brni, 3-bromo-7ni, 3-br-7-nitroindazole, tocris-0735, lopac-b-2050, d0u6zt, d0x5qj
PubChem CID 1649
IUPAC Name 3-bromo-7-nitro-2H-indazole
SMILES [O-][N+](=O)C1=CC=CC2=C(Br)NN=C12

Specifications

Color Yellow
Quantity 1 g
Sensitivity Light sensitive
Solubility Information Soluble in ethanol at 25mg/ml; Also soluble in DMSO
Formula Weight 242
Percent Purity ≥98%
Physical Form Powder
Chemical Name or Material 3-Bromo-7-nitroindazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.