missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-9-phenylcarbazole, 98%

Product Code. 15431319
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431319 100 g 100g
15411319 5 g 5g
15421319 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15431319 Supplier Thermo Scientific Alfa Aesar Supplier No. H64212.22
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1153-85-1 | C18H12BrN | 322.21 g/mol

Used in manufacturing of OLED materials. Also used as intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used in manufacturing of OLED materials. Also used as intermediate for pharmaceutical.

Solubility
Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1153-85-1
Molecular Formula C18H12BrN
Molecular Weight (g/mol) 322.21
MDL Number MFCD11977305
InChI Key KUBSCXXKQGDPPD-UHFFFAOYSA-N
Synonym 3-bromo-9-phenyl-9h-carbazole, 3-bromo-n-phenylcarbazole, 9h-carbazole, 3-bromo-9-phenyl, 3-bromo-9-phenyl-carbazole, ksc493s2d, 3-bromanyl-9-phenyl-carbazole, 3-bromo-9-phenyl-9,9a-dihydro-4ah-carbazole
PubChem CID 18942624
IUPAC Name 3-bromo-9-phenylcarbazole
SMILES BrC1=CC2=C(C=C1)N(C1=C2C=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 97°C to 98°C
Quantity 100 g
Beilstein 1315492
Solubility Information Slightly soluble in water.
Formula Weight 322.2
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 3-Bromo-9-phenylcarbazole

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.