missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-o-xylene, 99%

Product Code. p-7022906
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11412608

Brand: Thermo Scientific Alfa Aesar A15889.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 576-23-8 | C8H9Br | 185.06 g/mol

3-Bromo-o-xylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromo-o-xylene is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Not miscible or difficult to mix in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Containers which are opened must be carefully resealed and kept upright to prevent leakage. Store in cool place.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 576-23-8
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.06
MDL Number MFCD00000069
InChI Key WLPXNBYWDDYJTN-UHFFFAOYSA-N
Synonym 3-bromo-o-xylene, 3-bromo-1,2-dimethylbenzene, 2,3-dimethylbromobenzene, 2,3-dimethyl bromobenzene, benzene, bromodimethyl, 1-bromo-2,3-dimethyl-benzene, benzene, 1-bromo-2,3-dimethyl, bromoxilene, xylylbromid, bromo-o-xylene
PubChem CID 68472
IUPAC Name 1-bromo-2,3-dimethylbenzene
SMILES CC1=CC=CC(Br)=C1C

Specifications

Density 1.365
Boiling Point 212°C to 214°C
Quantity 5 g
Refractive Index 1.56
Beilstein 2077312
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 185.07
Percent Purity 99%
Chemical Name or Material 3-Bromo-o-xylene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.