missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoadamantane-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10475702
Change view
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10475702 1 g Amber Glass Bottle 1g
10394622 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10475702 Supplier Thermo Scientific Maybridge Supplier No. NRB01917DA
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 21816-08-0
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.143
MDL Number MFCD00167820
InChI Key DJUDQBVINJIMFO-UHFFFAOYSA-N
Synonym 1-bromo-3-adamantanecarboxylic acid, 3-bromo-1-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, 3-bromo, 3-bromo-adamantane-1-carboxylic acid, 3-adamantanecarboxylic acid, 1-bromo, 3-bromoadamantanecarboxylic acid, 3-bromotricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, acmc-1cnbw, maybridge1_005590, 5s,7r-3-bromoadamantane-1-carboxylate
PubChem CID 30818
IUPAC Name 3-bromoadamantane-1-carboxylic acid
SMILES C1C2CC3(CC1CC(C2)(C3)Br)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 259.14
Percent Purity 97%
Chemical Name or Material 3-Bromoadamantane-1-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.