missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzenesulfonyl chloride, 97%

Product Code. 11464493
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11464493 25 g 25g
11440433 1 g 1g
11454493 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11464493 Supplier Thermo Scientific Alfa Aesar Supplier No. A11507.14
 Restricted Item
Estimated Shipment: 04-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2905-24-0 | C6H4BrClO2S | 255.51 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2905-24-0
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00052313
InChI Key PJGOLCXVWIYXRQ-UHFFFAOYSA-N
Synonym 3-bromobenzene-1-sulfonyl chloride, 3-bromobenzenesulphonyl chloride, m-bromobenzenesulfonyl chloride, 3-bromobenzenesulfonylchloride, m-bromobenzenesulphonyl chloride, 3-bromo-benzenesulfonyl chloride, 3-bromophenyl chlorosulfone, 3-bromophenylsulfonyl chloride, buttpark 94\57-33, benzenesulfonyl chloride, 3-bromo
PubChem CID 17943
IUPAC Name 3-bromobenzenesulfonyl chloride
SMILES ClS(=O)(=O)C1=CC=CC(Br)=C1

Specifications

Melting Point 30°C to 33°C
Density 1.773
Boiling Point 90°C to 91°C (0.5 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.593
Quantity 25 g
UN Number UN3261
Beilstein 2691656
Sensitivity Moisture sensitive
Formula Weight 255.52
Percent Purity 97%
Chemical Name or Material 3-Bromobenzenesulfonyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.