missing translation for 'onlineSavingsMsg'
Learn More

3-Bromobenzo[b]thiophene, 95%

Product Code. 15425737
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15425737 10 g 10g
15435737 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15425737 Supplier Thermo Scientific Alfa Aesar Supplier No. H32948.09
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 7342-82-7 | C8H5BrS | 213.092 g/mol

Antifungal activity of synthetic di (hetero) arylamines based on the benzo [b] thiophene moiety.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Antifungal activity of synthetic di (hetero) arylamines based on the benzo [b] thiophene moiety.

Solubility
Sparingly Soluble (0.14 g/L) (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7342-82-7
Molecular Formula C8H5BrS
Molecular Weight (g/mol) 213.092
MDL Number MFCD00023009
InChI Key SRWDQSRTOOMPMO-UHFFFAOYSA-N
Synonym 3-bromobenzo b thiophene, 3-bromothianaphthene, 3-bromobenzothiophene, benzo b thiophene, 3-bromo, 3-bromo-benzo b thiophene, 3-bromo benzothiophene, benzo b thiophene, 3-bromo-,, zlchem 804
PubChem CID 123250
IUPAC Name 3-bromo-1-benzothiophene
SMILES C1=CC=C2C(=C1)C(=CS2)Br

Specifications

Density 1.629
Boiling Point 72°C to 74°C (0.27 mmHg)
Refractive Index 1.668
Quantity 10 g
Solubility Information Sparingly Soluble (0.14g/L) (25°C).
Formula Weight 213.1
Percent Purity 95%
Chemical Name or Material 3-Bromobenzo[b]thiophene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.