missing translation for 'onlineSavingsMsg'
Learn More

3-Bromochlorobenzene, 99%

Product Code. 10766561 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10766561 25 mL Glass bottle 25mL
10225580 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10766561 Supplier Thermo Scientific Acros Supplier No. 154200250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-37-2 | C6H4BrCl | 191.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108-37-2
Molecular Formula C6H4BrCl
Molecular Weight (g/mol) 191.45
MDL Number MFCD00000568
InChI Key JRGGUPZKKTVKOV-UHFFFAOYSA-N
Synonym 3-bromochlorobenzene, m-bromochlorobenzene, 3-chlorobromobenzene, benzene, 1-bromo-3-chloro, m-chlorobromobenzene, 1-chloro-3-bromobenzene, 3-chlorophenyl bromide, m-chlorophenyl bromide, m-bromophenyl chloride, 3-bromo-1-chlorobenzene
PubChem CID 7928
IUPAC Name 1-bromo-3-chlorobenzene
SMILES ClC1=CC=CC(Br)=C1

Specifications

Melting Point -21.5°C
CAS Min % 98.5
Color Colorless to Yellow
CAS Max % 100.0
Density 1.6300g/mL
Boiling Point 195.0°C to 197.0°C
Quantity 25 mL
Infrared Spectrum Authentic
Packaging Glass bottle
Physical Form Liquid
Refractive Index 1.5760 to 1.5780
Assay Percent Range 98.5% min. (GC)
Specific Gravity 1.63
Linear Formula BrC6H4Cl
Beilstein 05, 209
Solubility Information Solubility in water: insoluble
Formula Weight 191.45
Percent Purity 99%
Chemical Name or Material 3-Bromochlorobenzene, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3-Bromochlorobenzene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.