missing translation for 'onlineSavingsMsg'
Learn More

3-Bromocyclohexene, 95%

Product Code. 10627164 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 mL
50 mL
250 mL
Packaging:
Glass bottle
Unit Size:
10mL
250mL
50mL
This item is not returnable. View return policy

Product Code. 10627164

Brand: Thermo Scientific Acros 335122500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1521-51-3 | C6H9Br | 161.04 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1521-51-3
Molecular Formula C6H9Br
Molecular Weight (g/mol) 161.04
MDL Number MFCD00013775
InChI Key AJKDUJRRWLQXHM-UHFFFAOYSA-N
Synonym 3-bromocyclohex-1-ene, cyclohexene, 3-bromo, 1-bromo-2-cyclohexene, 3-bromo-1-cyclohexene, 1,2,3,4-tetrahydrobromobenzene, 3-bromo-cyclohexene, 2-cyclohexenyl bromide, 2-cyclohexen-1-yl bromide, 3-bromo cyclohexene, 3-bromanylcyclohexene
PubChem CID 137057
IUPAC Name 3-bromocyclohexene
SMILES C1CC=CC(C1)Br

Specifications

CAS Min % 94.0
CAS Max % 100.0
Color White
Density 1.3800g/mL
Boiling Point 64.0°C to 65.0°C (15.0 mmHg)
Flash Point 54°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Refractive Index 1.5280 to 1.5320
Quantity 250 mL
Specific Gravity 1.38
Formula Weight 161.04
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 3-Bromocyclohexene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Bromocyclohexene
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.