missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofluoranthene, TRC

Product Code. 30333723
Change view
Click to view available options
Quantity:
2.5 mg
5 mg
Unit Size:
2.5mg
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333723 5 mg 5mg
30329313 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30333723 Supplier Toronto Research Chemicals Supplier No. TRCB6948335MG
 Restricted Item
Estimated Shipment: 17-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromofluoranthene
CAS 13438-50-1
Molecular Formula C16 H9 Br
Quantity 5 mg
Synonym 3-Bromo-fluoranthene
InChI Formula InChI=1S/C16H9Br/c17-15-9-8-13-11-5-2-1-4-10(11)12-6-3-7-14(15)16(12)13/h1-9H
SMILES Brc1ccc2c3ccccc3c4cccc1c24
IUPAC Name 3-bromofluoranthene
Molecular Weight (g/mol) 281.147
Formula Weight 279.989
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.