missing translation for 'onlineSavingsMsg'
Learn More

3-Bromofuro[3,2-b]pyridine, 96%

Product Code. 15486497
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 15486497

Brand: Thermo Scientific Alfa Aesar H33652.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 92404-63-2 | C7H4BrNO | 198.019 g/mol

3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromofuro[3,2-b]pyridine is used as a starting material for synthesis of polyheterocycles.

Notes
Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92404-63-2
Molecular Formula C7H4BrNO
Molecular Weight (g/mol) 198.019
MDL Number MFCD19687763
InChI Key KHMAAFDMZSYVEF-UHFFFAOYSA-N
Synonym 3-bromofuro 3,2-b pyridine, furo 3,2-b pyridine, 3-bromo, acmc-20lvu1
PubChem CID 641477
IUPAC Name 3-bromofuro[3,2-b]pyridine
SMILES C1=CC2=C(C(=CO2)Br)N=C1

Specifications

Melting Point 51°C to 53°C
Quantity 1 g
Formula Weight 198.02
Percent Purity 96%
Chemical Name or Material 3-Bromofuro[3,2-b]pyridine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.