missing translation for 'onlineSavingsMsg'
Learn More

3-Bromomethyl-2(1H)-quinoxalinone, tech. 90%

Product Code. 11486310
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11486310 5 g 5g
11496310 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11486310 Supplier Thermo Scientific Alfa Aesar Supplier No. H27169.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 62235-61-4 | C9H7BrN2O | 239.072 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 62235-61-4
Molecular Formula C9H7BrN2O
Molecular Weight (g/mol) 239.072
MDL Number MFCD02660613
InChI Key SOTAKCFOTFCVSS-UHFFFAOYSA-N
Synonym 3-bromomethyl quinoxalin-2-ol, 3-bromomethylquinoxalin-2-one, 3-bromomethyl-1h-quinoxalin-2-one, 3-bromomethyl-2-quinoxalinol, 3-bromomethyl-2 1h-quinoxalinone, 2 1h-quinoxalinone, 3-bromomethyl, 3-bromomethyl hydroquinoxalin-2-one, acmc-20agnv, 3-bromomethyl quinoxaline
PubChem CID 286254
IUPAC Name 3-(bromomethyl)-1H-quinoxalin-2-one
SMILES C1=CC=C2C(=C1)NC(=O)C(=N2)CBr

Specifications

Melting Point >300°C (decomposition)
Quantity 5 g
UN Number UN1759
Formula Weight 239.07
Percent Purity 90%
Grade Technical
Chemical Name or Material 3-Bromomethyl-2(1H)-quinoxalinone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.