missing translation for 'onlineSavingsMsg'
Learn More

3-Bromomethyl-3-oxetanemethanol, 95%

Product Code. 15423888
Change view
Click to view available options
Quantity:
250 mg
500 mg
Unit Size:
250mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15423888 250 mg 250mg
15433888 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15423888 Supplier Thermo Scientific Alfa Aesar Supplier No. H57413.MD
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 22633-44-9 | C5H9BrO2 | 181.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22633-44-9
Molecular Formula C5H9BrO2
Molecular Weight (g/mol) 181.03
MDL Number MFCD09800598
InChI Key SESXZSLSTRITGO-UHFFFAOYSA-N
PubChem CID 529264
IUPAC Name [3-(bromomethyl)oxetan-3-yl]methanol
SMILES OCC1(CBr)COC1

Specifications

Density 1.598
Boiling Point 242°C
Flash Point 100°C (212°F)
Quantity 250 mg
Synonym 3-bromomethyl oxetan-3-yl methanol, 3-bromomethyl-3-oxetanemethanol, 3-bromomethyl-3-hydroxymethyloxetane, 3-oxetanemethanol, 3-bromomethyl, oxetane, 3-bromomethyl-3-hydroxymethyl, 3-bromomethyl-3-hydroxymethyl oxetane, 3-bromomethyl oxetan-3-yl methan-1-ol, 3-oxetanemethanol,3-bromomethyl, 2-bromomethyl-2-hydroxymethyl-1,3-epoxypropane
Formula Weight 181.03
Percent Purity 95%
Chemical Name or Material 3-Bromomethyl-3-oxetanemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.