missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)-5-chlorobenzo[b]thiophene, TRC

Product Code. 30346780
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30346780 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30346780 Supplier Toronto Research Chemicals Supplier No. TRCB697123100MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Bromomethyl)-5-chlorobenzo[b]thiophene
CAS 1198-51-2
Molecular Formula C9 H6 Br Cl S
Quantity 100 mg
InChI Formula InChI=1S/C9H6BrClS/c10-4-6-5-12-9-2-1-7(11)3-8(6)9/h1-3,5H,4H2
SMILES Clc1ccc2scc(CBr)c2c1
IUPAC Name 3-(bromomethyl)-5-chloro-1-benzothiophene
Molecular Weight (g/mol) 261.566
Formula Weight 259.906
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.