missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)-5-methylisoxazole, 97%, Thermo Scientific™

Product Code. 10766075
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10766075 5 g Amber Glass Bottle 5g
10497032 250 mg Amber Glass Bottle 250mg
10542472 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10766075 Supplier Thermo Scientific Maybridge Supplier No. CC10408DE
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 130628-75-0
Molecular Formula C5H6BrNO
Molecular Weight (g/mol) 176.01
MDL Number MFCD02677720
InChI Key ASGJFGPILHALRC-UHFFFAOYSA-N
Synonym 3-bromomethyl-5-methylisoxazole, 3-bromomethyl-5-methyl-1,2-oxazole, 3-bromomethyl-5-methyl-isoxazole, isoxazole,3-bromomethyl-5-methyl, zlchem 1313, acmc-1ca86, 3-bromomethyl-5-methylisoxazol, 5-brimomethyl-3-methylisoxazle
PubChem CID 2776304
IUPAC Name 3-(bromomethyl)-5-methyl-1,2-oxazole
SMILES CC1=CC(CBr)=NO1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 176.01
Percent Purity 97%
Chemical Name or Material 3-(Bromomethyl)-5-methylisoxazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.