missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-(Bromomethyl)pyridine hydrobromide, 97%
Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packungsgröße:
1g
5g
Beschreibung
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spezifikation
Spezifikation
| Melting Point | 150.0°C to 155.0°C |
| Assay Percent Range | 97% |
| Packaging | Glass bottle |
| Quantity | 5 g |
| Formula Weight | 252.93 |
| Percent Purity | 97% |
| Chemical Name or Material | 3-(Bromomethyl)pyridine hydrobromide |
Sicherheit und Handhabung
Produktidentifikator
- 3-(Bromomethyl)pyridine hydrobromide
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 B
- H318-Causes serious eye damage.
- H314-Causes severe skin burns and eye damage.
- H312-Harmful in contact with skin.
- H332-Harmful if inhaled.
- H302-Harmful if swallowed.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur