missing translation for 'onlineSavingsMsg'
Learn More

3-(Bromomethyl)pyridine hydrobromide, TRC

Product Code. 30338052
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
Product Code. Quantity unitSize
30338052 10 g 10g
1 options
This item is not returnable. View return policy

Product Code. 30338052

Brand: Toronto Research Chemicals TRCB06902010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-(Bromomethyl)pyridine hydrobromide
CAS 4916-55-6
Molecular Formula C6H6BrN . HBr
Quantity 10 g
InChI Formula InChI=1S/C6H6BrN.BrH/c7-4-6-2-1-3-8-5-6;/h1-3,5H,4H2;1H
SMILES Br.BrCc1cccnc1
IUPAC Name 3-(bromomethyl)pyridine;hydrobromide
Molecular Weight (g/mol) 252.93
Formula Weight 250.8945
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.