missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromophenylhydrazine Hydrochloride, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | 3-Bromophenylhydrazine Hydrochloride |
| CAS | 27246-81-7 |
| Molecular Formula | C6 H7 Br N2 . Cl H |
| Quantity | 10 g |
| Synonym | 1-(3-Bromophenyl)hydrazine Hydrochloride, m-Bromphenylhydrazine Hydrochloride |
| InChI Formula | InChI=1S/C6H7BrN2.ClH/c7-5-2-1-3-6(4-5)9-8;/h1-4,9H,8H2;1H |
| SMILES | Cl.NNc1cccc(Br)c1 |
| IUPAC Name | (3-bromophenyl)hydrazine;hydrochloride |
| Molecular Weight (g/mol) | 223.5 |
| Formula Weight | 221.9559 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Ser du en mulighed for forbedring?Del en indholdskorrektion