missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropionitrile, 97%

Product Code. 11446896
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11446896 100 g 100g
11436896 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11446896 Supplier Thermo Scientific Alfa Aesar Supplier No. B23998.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2417-90-5 | C3H4BrN | 133.976 g/mol

3-Bromopropionitrile was used as the initiator in the atom transfer radical polymerization of acrylonitrile. It was used in coupling reactions of a number of aromatic and heteroaromatic phenols with alkyl, acyl or benzoyl halides in acetonitrile with cesium fluoride-Celite.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopropionitrile was used as the initiator in the atom transfer radical polymerization of acrylonitrile. It was used in coupling reactions of a number of aromatic and heteroaromatic phenols with alkyl, acyl or benzoyl halides in acetonitrile with cesium fluoride-Celite.

Solubility
Soluble in ethanol, ether.

Notes
Store in a cool, dry place in tightly closed container. Store at room temperature. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2417-90-5
Molecular Formula C3H4BrN
Molecular Weight (g/mol) 133.976
MDL Number MFCD00001951
InChI Key CQZIEDXCLQOOEH-UHFFFAOYSA-N
Synonym 3-bromopropionitrile, propanenitrile, 3-bromo, 2-bromoethyl cyanide, 3-bromopropiononitrile, 1-bromo-2-cyanoethane, beta-bromopropionitrile, 2-cyanoethyl bromide, propionitrile, 3-bromo, usaf do-51, beta-bromopriopionitrile
PubChem CID 17020
IUPAC Name 3-bromopropanenitrile
SMILES C(CBr)C#N

Specifications

Density 1.615
Boiling Point 200°C
Flash Point 97°C (206°F)
Assay Percent Range 0.98
Refractive Index 1.481
Quantity 100 g
UN Number UN3276
Beilstein 1738565
Solubility Information Soluble in ethanol,ether.
Formula Weight 133.98
Percent Purity 98%
Chemical Name or Material 3-Bromopropionitrile
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.