missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropylamine hydrobromide, 98%

Product Code. 11440857
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
Product Code. Quantity unitSize
11440857 25 g 25g
11450857 100 g 100g
2 options
This item is not returnable. View return policy

Product Code. 11440857

Brand: Thermo Scientific Alfa Aesar B23254.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5003-71-4 | C3H9Br2N | 218.92 g/mol

3-Bromopropylamine hydrobromide is used as pharmaceutical raw materials and intermediates. Homotaurine is synthesized from 3-bromopropylamine hydrobromide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromopropylamine hydrobromide is used as pharmaceutical raw materials and intermediates. Homotaurine is synthesized from 3-bromopropylamine hydrobromide.

Solubility
Soluble in water (50 mg/ml).

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5003-71-4
Molecular Formula C3H9Br2N
Molecular Weight (g/mol) 218.92
MDL Number MFCD00012912
InChI Key PQIYSSSTRHVOBW-UHFFFAOYSA-N
Synonym 3-bromopropylamine hydrobromide, 3-bromopropan-1-amine hydrobromide, 3-aminopropyl bromide hydrobromide, 1-propanamine, 3-bromo-, hydrobromide, 3-bromopropylammonium bromide, 3-bromopropanamine hydrobromide, 1-amino-3-bromopropane hydrobromide, 3-bromo-1-propanamine, hydrobromide, 3-brominepropylaminehydrobromide, 3-bromopropylamine, hydrobromide
PubChem CID 78701
IUPAC Name 3-bromopropan-1-amine;hydrobromide
SMILES C(CN)CBr.Br

Specifications

Melting Point 172°C to 174°C
Quantity 25 g
Beilstein 3906418
Sensitivity Hygroscopic
Solubility Information Soluble in water (50mg/ml).
Formula Weight 218.93
Percent Purity 98%
Chemical Name or Material 3-Bromopropylamine hydrobromide
compliance-icons
Product Identifier
  • 3-Bromopropylamine hydrobromide
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3-Bromopropylamine hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.