missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Bromopropylboronic acid pinacol ester, 98%
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Description
Used as primary and secondary intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Used as primary and secondary intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Used as primary and secondary intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Density | 1.177 |
| Boiling Point | 61°C to 64°C (0.35 mmHg) |
| Quantity | 5 g |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 248.96 |
| Percent Purity | 98% |
| Chemical Name or Material | 3-Bromopropylboronic acid pinacol ester |
Safety and Handling
Product Identifier
- 3-Bromopropylboronic acid pinacol ester
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction