missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopropylboronic acid pinacol ester, 98%

Product Code. 15493967
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15493967 1 g 1g
15403977 5 g 5g
15413977 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15493967 Supplier Thermo Scientific Alfa Aesar Supplier No. H30646.03
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 124215-44-7 | C9H18BBrO2 | 248.96 g/mol

Used as primary and secondary intermediate for pharmaceutical.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as primary and secondary intermediate for pharmaceutical.

Solubility
Not miscible or difficult to mix with water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124215-44-7
Molecular Formula C9H18BBrO2
Molecular Weight (g/mol) 248.96
MDL Number MFCD10567053
InChI Key CHQXFJUKMDJWHO-UHFFFAOYSA-N
Synonym 2-3-bromopropyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3-bromopropylboronic acid pinacol ester, 1,3,2-dioxaborolane,2-3-bromopropyl-4,4,5,5-tetramethyl, acmc-1c5j4, 3-bromopropylboronic acid, pinacol ester, 2-3-bromopropyl-4?4?5?5-tetramethyl-1?3?2-dioxaborolane
PubChem CID 14836669
IUPAC Name 2-(3-bromopropyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane
SMILES CC1(C)OB(CCCBr)OC1(C)C

Specifications

Density 1.177
Boiling Point 61°C to 64°C (0.35 mmHg)
Quantity 1 g
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 248.96
Percent Purity 98%
Chemical Name or Material 3-Bromopropylboronic acid pinacol ester
compliance-icons
Product Identifier
  • 3-Bromopropylboronic acid pinacol ester
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 3-Bromopropylboronicacid pinacol ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.