missing translation for 'onlineSavingsMsg'
Learn More

3-Bromopyridine, TRC

Product Code. 30340218
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30340218 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30340218 Supplier Toronto Research Chemicals Supplier No. TRCB6867402.5G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifikationer

Chemical Name or Material 3-Bromopyridine
CAS 626-55-1
Molecular Formula C5H4BrN
Quantity 2.5 g
Synonym 3-Pyridyl Bromide, NSC 3974, Pyridin-3-yl Bromide, m-Bromopyridine
InChI Formula InChI=1S/C5H4BrN/c6-5-2-1-3-7-4-5/h1-4H
SMILES Brc1cccnc1
IUPAC Name 3-bromopyridine
Molecular Weight (g/mol) 158
Formula Weight 156.9527
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.