missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ 3-Bromopyruvate

Product Code. 18758036
Change view
Click to view available options
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
18758036 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18758036 Supplier R&D Systems™ Supplier No. 7512/100
  • ONLINE EXCLUSIVE PRICE
    113.57 €
    102.90 € / 100mg
    Save 10.67 €
    9% Off
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. MCT1 inhibitor; also inhibits hexokinase II

3-Bromopyruvate is a monocarboxylate transporter (MCT1) inhibitor (IC50 = 14.7 μM, 24.2 μM and 28.6 μM in SiHa, UM-UC-3, and HCT-116 human cancer cell lines, respectively). 3-Bromopyruvate shows antitumor activity in vitro and in vivo. 3-Bromopyruvate is also an inhibitor of hexokinase II, and decreases apoptosis in dopaminergic neurons in Parkinson's disease models in vitro and in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Bromo-2-oxopropanoic acid
CAS 1113-59-3
Quantity 100 mg
Product Line Tocris
Target Monocarboxylate Transporter Inhibitors
Molecular Formula C3H3BrO3
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.