missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carbaldehyde, 97%, Thermo Scientific™

Product Code. 10293802
Change view
Click to view available options
Quantity:
1 g
25 g
Unit Size:
1g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10293802 1 g 1g
10142754 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10293802 Supplier Thermo Scientific Maybridge Supplier No. SEW01928DA
 Restricted Item
Estimated Shipment: 28-10-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 930-96-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00126680
InChI Key BCZHCWCOQDRYGS-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-carboxaldehyde, 3-bromo-2-formylthiophene, 3-bromo-2-thiophenecarbaldehyde, 3-bromo-2-thiophene carboxaldehyde, 3-bromo-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 3-bromo, bromothiophenecarbaldehyde, 3-bromothiophene-2-aldehyde, pubchem5327, acmc-209rjz
PubChem CID 2797079
IUPAC Name 3-bromothiophene-2-carbaldehyde
SMILES BrC1=C(SC=C1)C=O

Specifications

CAS Min % 97
Density 1.81g/mL
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 191.05
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.