missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromothiophene-2-carboxaldehyde, 94%

Product Code. 11398666
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11398666 5 g 5g
11388666 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11398666 Supplier Alfa Aesar™ Supplier No. L19386.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
3-Bromothiophene-2-carboxaldehyde is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Not miscible in water.

Notes
Air Sensitive. Store away from air. Store under dry inert gas. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-96-1
Molecular Formula C5H3BrOS
Molecular Weight (g/mol) 191.04
MDL Number MFCD00126680
InChI Key BCZHCWCOQDRYGS-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-carboxaldehyde, 3-bromo-2-formylthiophene, 3-bromo-2-thiophenecarbaldehyde, 3-bromo-2-thiophene carboxaldehyde, 3-bromo-thiophene-2-carbaldehyde, 2-thiophenecarboxaldehyde, 3-bromo, bromothiophenecarbaldehyde, 3-bromothiophene-2-aldehyde, pubchem5327, acmc-209rjz
PubChem CID 2797079
IUPAC Name 3-bromothiophene-2-carbaldehyde
SMILES BrC1=C(SC=C1)C=O

Specifications

Boiling Point 110°C to 113°C (10 mmHg)
Quantity 5 g
Beilstein 109757
Formula Weight 191.05
Percent Purity 94%
Chemical Name or Material 3-Bromothiophene-2-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.