missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carboxamide, 99%, Thermo Scientific Chemicals

Product Code. 11312907
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11312907 250 mg 250mg
11302907 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11312907 Supplier Thermo Scientific Alfa Aesar Supplier No. L16138.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Alternative Produkte anzeigen

missing translation for 'validMainframeMsg'

Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

CAS: 78031-18-2 | Molecular Formula: C5H4BrNOS | Molecular Weight: 206.06 g/mol

3-Bromothiophene-2-carboxamide is used in Stille cross-coupling conditions were used to couple the resin-bound peptide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Bromothiophene-2-carboxamide is used in Stille cross-coupling conditions were used to couple the resin-bound peptide.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemische Identifikatoren

CAS 78031-18-2
Molecular Formula C5H4BrNOS
Molecular Weight (g/mol) 206.06
MDL Number MFCD00173835
InChI Key KWXUWNXTRUZHNQ-UHFFFAOYSA-N
Synonym maybridge1_007895, 3-bromo-2-carboxamidothiophene, 2-thiophenecarboxamide,3-bromo, 3-bromo-thiophene-2-carboxamide, 3-bromo-2-thiophenecarboxamide #, buttpark 180\02-08, 3-bromo thiophene-2-carboxamide
PubChem CID 605721
IUPAC Name 3-bromothiophene-2-carboxamide
SMILES NC(=O)C1=C(Br)C=CS1

Spezifikation

Melting Point 106°C to 108°C
Quantity 250 mg
Beilstein 4937660
Solubility Information Insoluble in water.
Formula Weight 206.06
Percent Purity 99%
Chemical Name or Material 3-Bromothiophene-2-carboxamide

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.