missing translation for 'onlineSavingsMsg'
Learn More

3-Bromothiophene-2-carboxylic acid, 97%

Product Code. 11463924
Change view
Click to view available options
Quantity:
0.25 g
1 g
5 g
Unit Size:
0.25g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11463924 1 g 1g
11483924 0.25 g 0.25g
11473924 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11463924 Supplier Thermo Scientific Alfa Aesar Supplier No. A11935.03
 Restricted Item
Estimated Shipment: 22-07-2026
Add to basket
This item is not returnable. View return policy

CAS: 7311-64-0 | C5H3BrO2S | 207.041 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7311-64-0
Molecular Formula C5H3BrO2S
Molecular Weight (g/mol) 207.041
MDL Number MFCD00052291
InChI Key VQQLWBUTTMNMFT-UHFFFAOYSA-N
Synonym 3-bromo-2-thiophenecarboxylic acid, 3-bromo-2-thenoic acid, 2-thiophenecarboxylic acid, 3-bromo, 3-bromo-thiophene-2-carboxylic acid, maybridge1_007344, 3-bromothiophene-2-carboxylicacid, bromothiophenecarboxylic, pubchem10096, ksc494c9j, bromo-thiophene-2-carboxylic acid
PubChem CID 2739688
IUPAC Name 3-bromothiophene-2-carboxylic acid
SMILES C1=CSC(=C1Br)C(=O)O

Specifications

Melting Point 195°C to 196°C
Quantity 1 g
Beilstein 121585
Formula Weight 207.05
Percent Purity 97%
Chemical Name or Material 3-Bromothiophene-2-carboxylic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.