missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-Bromothiophene-2-sulfonyl chloride, 90+%

Product Code. 15455967
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15455967 250 mg 250mg
15445967 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15455967 Supplier Alfa Aesar™ Supplier No. H33170.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 170727-02-3
Molecular Formula C4H2BrClO2S2
Molecular Weight (g/mol) 261.532
MDL Number MFCD08059484
InChI Key OPLMNCCLQDVCTP-UHFFFAOYSA-N
Synonym 3-bromothiophene-2-sulfonylchloride, 2-thiophenesulfonylchloride, 3-bromo, 3-bromo-thiophene-2-sulfonyl chloride, 3-bromo-thiophene-2-sulphonyl chloride, 3-bromo 2-thienyl chlorosulfone, 3-bromo-2-thiophenesulfonyl chloride, 3-bromothiophene-2-sulphonyl chloride, 2-thiophenesulfonyl chloride, 3-bromo
PubChem CID 13675598
IUPAC Name 3-bromothiophene-2-sulfonyl chloride
SMILES C1=CSC(=C1Br)S(=O)(=O)Cl

Specifications

Melting Point 39°C to 41°C
Quantity 250 mg
UN Number UN3261
Formula Weight 261.55
Percent Purity ≥90%
Chemical Name or Material 3-Bromothiophene-2-sulfonyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.