missing translation for 'onlineSavingsMsg'
Learn More

3-Buten-2-ol, 97%

Product Code. 10260223 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10260223 25 mL Glass bottle 25mL
10107193 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10260223 Supplier Thermo Scientific Acros Supplier No. 387070250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 598-32-3 | C4H8O | 72.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 598-32-3
Molecular Formula C4H8O
Molecular Weight (g/mol) 72.11
MDL Number MFCD00004543
InChI Key MKUWVMRNQOOSAT-UHFFFAOYNA-N
Synonym 3-buten-2-ol, 1-buten-3-ol, methyl vinylcarbinol, 3-butene-2-ol, 3-hydroxy-1-butene, propenol, 1-methyl, methyl vinyl carbinol, 1-methyl-2-propenol, 2-methyl-2-propenol, 1-methylallyl alcohol
PubChem CID 11716
IUPAC Name but-3-en-2-ol
SMILES CC(O)C=C

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -100.0°C
Color Colorless
Density 0.8300g/mL
Boiling Point 96.0°C to 98.0°C
Flash Point 16°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula H2C=CHCH(OH)CH3
Refractive Index 1.4130 to 1.4150
Quantity 25 mL
Specific Gravity 0.83
Solubility Information Solubility in water: soluble
Formula Weight 72.11
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Buten-2-ol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Buten-2-ol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and any thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.