missing translation for 'onlineSavingsMsg'
Learn More

3-Butyn-1-ol, 97%

Product Code. 10655592 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 mL
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
5mL
This item is not returnable. View return policy

Product Code. 10655592

Brand: Thermo Scientific Acros 157330250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 927-74-2 | C4H6O | 70.09 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 927-74-2
Molecular Formula C4H6O
Molecular Weight (g/mol) 70.09
MDL Number MFCD00002955
InChI Key OTJZCIYGRUNXTP-UHFFFAOYSA-N
Synonym 3-butyn-1-ol, 3-butynol, 3-butyne-1-ol, 4-hydroxy-1-butyne, 3-butynyl alcohol, 1-butyn-4-ol, 2-hydroxyethylacetylene, homopropargyl alcohol, 2-hydroxyethyl acetylene, 4-hydroxy-butyne
PubChem CID 13566
ChEBI CHEBI:27444
IUPAC Name but-3-yn-1-ol
SMILES C#CCCO

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point -63.6°C
Color Colorless to Yellow
Density 0.9260g/mL
Boiling Point 128.8°C
Flash Point 36°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HC≡CCH2CH2OH
Refractive Index 1.4397 to 1.4417
Quantity 25 mL
Beilstein 01, IV, 2219
Specific Gravity 0.926
Solubility Information Solubility in water: miscible
Formula Weight 70.09
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 3-Butyn-1-ol, 97%
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 3-Butyn-1-ol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Corrección del contenido de un producto

Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.

Título del producto

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.

Gracias por ayudarnos a mejorar nuestra web. Su comentario ha sido enviado