missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxy-4-methoxybenzeneboronic acid, 98%, Thermo Scientific Chemicals

Product Code. 15479777
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479777 1 g 1g
15489777 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15479777 Leverandør Thermo Scientific Alfa Aesar Leverandørnr. H53005.03
 Restricted Item
Estimated Shipment: 14-12-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 913836-12-1 | Molecular Formula: C8H9BO5 | Molecular Weight: 195.965 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 913836-12-1
Molecular Formula C8H9BO5
Molecular Weight (g/mol) 195.965
MDL Number MFCD06203322
InChI Key YZKWFWNYFKBAHO-UHFFFAOYSA-N
Synonym 3-carboxy-4-methoxyphenylboronic acid, 5-dihydroxyboranyl-2-methoxybenzoic acid, benzoicacid, 5-borono-2-methoxy-9ci, 3-carboxy-4-methoxybenzeneboronic acid, benzoic acid, 5-borono-2-methoxy, acmc-209raw, ksc486i0n, 5-dihydroxyboryl-2-methoxybenzoic acid
PubChem CID 44119154
IUPAC Name 5-borono-2-methoxybenzoic acid
SMILES B(C1=CC(=C(C=C1)OC)C(=O)O)(O)O

Tekniske data

Melting Point 176°C to 177°C
Quantity 1 g
Formula Weight 195.97
Percent Purity 98%
Chemical Name or Material 3-Carboxy-4-methoxybenzeneboronic acid

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.