missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxybenzaldehyde, 97%

Product Code. 11409177
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11409177

Brand: Thermo Scientific Alfa Aesar B21277.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 619-21-6 | C8H6O3 | 150.133 g/mol

3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Formylbenzoic acid is used in the synthesis of bicyclic cis-2-azetidinone derivatives via Ugi 4-centre 3-component reaction, porphyrin capped with a steroidal superstructure bearing convergent hydroxy groups and 3-[(4-amino-1,2-dihydro-1-oxo-2-phenyl-1,2,4-triazolo[4,3-a]quinoxalin-6-yl)amino]methyl benzoic acid.

Solubility
Soluble in methanol (100 mg/mL) gives a clear to slightly hazy, colorless to very faintly brown.

Notes
Air sensitive. Store away from oxidizing agents, reducing agents and bases. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 619-21-6
Molecular Formula C8H6O3
Molecular Weight (g/mol) 150.133
MDL Number MFCD00039575
InChI Key UHDNUPHSDMOGCR-UHFFFAOYSA-N
Synonym 3-carboxybenzaldehyde, benzoic acid, 3-formyl, m-formyl benzoic acid, 3-formyl benzoic acid, 3-formyl-benzoic acid, unii-hb95grm95d, m-carboxybenzaldehyde, hb95grm95d, 3-formylbenzoicacid, isophthalaldehydsaure
PubChem CID 12077
IUPAC Name 3-formylbenzoic acid
SMILES C1=CC(=CC(=C1)C(=O)O)C=O

Specifications

Melting Point 173°C to 175°C
Quantity 1 g
Beilstein 2206413
Solubility Information Soluble in methanol (100mg/mL) gives a clear to slightly hazy,colorless to very faintly brown.
Formula Weight 150.13
Percent Purity 97%
Chemical Name or Material 3-Carboxybenzaldehyde

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.