missing translation for 'onlineSavingsMsg'
Learn More

3-Carboxyphenylboronic Acid, TRC

Product Code. 30367844
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367844 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30367844 Supplier Toronto Research Chemicals Supplier No. TRCC1813832.5G
 Restricted Item
Estimated Shipment: 19-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 3-Carboxyphenylboronic Acid
CAS 25487-66-5
Molecular Formula C7H7BO4
Quantity 2.5 g
Synonym 3-Boronobenzoic Acid, 3-(Dihydroxyborane)benzoic Acid, 3-(Dihydroxyboranyl)benzoic Acid, 3-(Dihydroxyboryl)benzoic Acid, 3-Carboxybenzeneboronic Acid, 3-Carboxyphenylboronic Acid, m-Carboxyphenylboronic Acid
InChI Formula InChI=1S/C7H7BO4/c9-7(10)5-2-1-3-6(4-5)8(11)12/h1-4,11-12H,(H,9,10)
SMILES OB(O)c1cccc(c1)C(=O)O
IUPAC Name 3-boronobenzoic acid
Molecular Weight (g/mol) 165.94
Formula Weight 166.0437
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.